2-(2-Bromo-4-methoxyphenoxy)acetonitrile 98%

Supplier: Ambeed
Warning

A421728-1G A421728-250MG A421728-5G
77016-464EA 9 USD
77016-464 77016-466 77016-468
2-(2-Bromo-4-methoxyphenoxy)acetonitrile 98%
2-(2-Bromo-4-methoxyphenoxy)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO₂
MDL Number: MFCD09906448
CAS Number: 951918-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR