4-Morpholino-2-pyridinamine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-(morpholin-4-yl)pyridine, 4-Morpholinopyridin-2-amine

A421344-100MG A421344-250MG
77004-110EA 40 USD
77004-110 77004-112
4-Morpholino-2-pyridinamine 95%
4-Morpholino-2-pyridinamine

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O
MW: 179.2200 g/mol
MDL Number: MFCD09991750
CAS Number: 722549-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR