(S)-Ethyl 2-((tert-butyldimethylsilyl)oxy)propanoate 95%

Supplier: Ambeed
Warning

A421105-100G A421105-25G A421105-5G
77016-458EA 560.16 USD
77016-458 77016-460 77016-462
(S)-Ethyl 2-((tert-butyldimethylsilyl)oxy)propanoate 95%
(S)-Ethyl 2-((tert-butyldimethylsilyl)oxy)propanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₄O₃Si
MW: 232.39 g/mol
MDL Number: MFCD00192135
CAS Number: 106513-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR