Ethyl-4-[(tert-butoxycarbonyl)amino]-3-oxobutanoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-((tert-butoxycarbonyl)amino)-3-oxobutanoate

A420965-250MG A420965-1G A420965-5G
76990-446EA 35.01 USD
76990-446 77572-312 77594-296
Ethyl-4-[(tert-butoxycarbonyl)amino]-3-oxobutanoate 95%
Ethyl-4-[(tert-butoxycarbonyl)amino]-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
MDL Number: MFCD11616704
CAS Number: 67706-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR