9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester 97%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dimethylfluorene-2,7-diboronic acid Bis(pinacol) ester, 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dimethylfluorene, 2,2'-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

A420792-1G A420792-250MG A420792-25G A420792-5G
77004-104EA 105.03 USD
77004-104 77004-106 77004-108 77882-096 77882-097
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester 97%
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester

Precaución: for Research Use Only

Formula: C₂₇H₃₆B₂O₄
MW: 446.20 g/mol
MDL Number: MFCD08704230
CAS Number: 325129-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR