Ethyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate

A420565-1G A420565-250MG A420565-5G
76990-440EA 22.28 USD
76990-440 76990-442 76990-444
Ethyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.2300 g/mol
MDL Number: MFCD00139509
CAS Number: 2549-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR