Methyl 3-bromo-4-ethoxybenzoate 95%

Supplier: Ambeed
Warning

A420357-100MG A420357-1G A420357-250MG A420357-5G
76990-432EA 20.16 USD
76990-432 76990-434 76990-436 76990-438
Methyl 3-bromo-4-ethoxybenzoate 95%
Methyl 3-bromo-4-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.09 g/mol
MDL Number: MFCD06801000
CAS Number: 24507-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR