Fmoc-Tyr(3,5-I2)-OH 98%

Supplier: Ambeed
Warning

A420276-10G A420276-1G A420276-5G A420276-25G
77033-862EA 89 USD
77033-862 77033-864 77033-866 77673-032
Fmoc-Tyr(3,5-I2)-OH 98%
Fmoc-Tyr(3,5-I2)-OH

Precaución: for Research Use Only

Formula: C₂₄H₁₉I₂NO₅
MDL Number: MFCD00063330
CAS Number: 103213-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR