Methyl-3-{[(2-methoxy-2-oxoethyl)amino]sulfonyl}thiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(N-(2-methoxy-2-oxoethyl)sulphamoyl)thiophene-2-carboxylate

A419850-100G A419850-25G A419850-5G
77004-096EA 102.91 USD
77004-096 77004-098 77004-100
Methyl-3-{[(2-methoxy-2-oxoethyl)amino]sulfonyl}thiophene-2-carboxylate 98%
Methyl-3-{[(2-methoxy-2-oxoethyl)amino]sulfonyl}thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₆S₂
MW: 293.32 g/mol
MDL Number: MFCD01570165
CAS Number: 106820-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR