3-Bromo-6-fluoro-1H-indazole 95%

Supplier: Ambeed
Warning

A419625-250MG A419625-1G
77033-852EA 26.53 USD
77033-852 77673-030
3-Bromo-6-fluoro-1H-indazole 95%
3-Bromo-6-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
MDL Number: MFCD07781603
CAS Number: 885522-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR