2-Bromo-1,3-dimethyl-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A419243-1G A419243-25G A419243-5G
77016-438EA 18 USD
77016-438 77016-440 77016-442
2-Bromo-1,3-dimethyl-5-nitrobenzene 95%
2-Bromo-1,3-dimethyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MDL Number: MFCD02178896
CAS Number: 53906-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR