(5-Chloro-1-benzofuran-2-yl)(4-methoxyphenyl)methanone 95%

Supplier: Ambeed
Warning

Synonyms: (5-Chlorobenzofuran-2-yl)(4-methoxyphenyl)methanone

A419100-1G A419100-250MG
77004-088EA 109.28 USD
77004-088 77004-090
(5-Chloro-1-benzofuran-2-yl)(4-methoxyphenyl)methanone 95%
(5-Chloro-1-benzofuran-2-yl)(4-methoxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₁₆H₁₁ClO₃
MW: 286.72 g/mol
MDL Number: MFCD12922712
CAS Number: 220526-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR