tert-Butyl 3-bromo-5-fluoro-1H-indazole-1-carboxylate 97%

Supplier: Ambeed
Warning

A419091-100MG A419091-1G A419091-250MG
76990-412EA 21.22 USD
76990-412 76990-414 76990-416
tert-Butyl 3-bromo-5-fluoro-1H-indazole-1-carboxylate 97%
tert-Butyl 3-bromo-5-fluoro-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrFN₂O₂
MW: 315.14 g/mol
MDL Number: MFCD06739131
CAS Number: 885271-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR