tert-Butyl 1H-indol-5-ylcarbamate 97%

Supplier: Ambeed
Warning

A419007-10G A419007-1G A419007-250MG A419007-5G
77016-430EA 572.89 USD
77016-430 77016-432 77016-434 77016-436
tert-Butyl 1H-indol-5-ylcarbamate 97%
tert-Butyl 1H-indol-5-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MW: 232.27 g/mol
MDL Number: MFCD04114771
CAS Number: 184031-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR