4-Bromophenyl trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Bromophenyl triflate
A418962-1G
A418962-5G
77033-832EA
25
USD
77033-832
77033-834
4-Bromophenyl trifluoromethanesulfonate 98%
4-Bromophenyl trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O₃S MW: 305.069 g/mol |
MDL Number:
MFCD07784325 CAS Number: 66107-30-0 UN: 3082 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...