3-Iodo-4-(piperidin-1-ylmethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A418756-100MG A418756-250MG
76990-408EA 55.17 USD
76990-408 76990-410
3-Iodo-4-(piperidin-1-ylmethyl)benzoic acid 95%
3-Iodo-4-(piperidin-1-ylmethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆INO₂
MW: 345.17 g/mol
MDL Number: MFCD11110810
CAS Number: 1131614-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR