2-(4-Bromophenyl)butanoic acid 97%

Supplier: Ambeed
Warning

A418751-100MG A418751-250MG A418751-5G
77004-082EA 18.04 USD
77004-082 77004-084 77004-086
2-(4-Bromophenyl)butanoic acid 97%
2-(4-Bromophenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD01737245
CAS Number: 99070-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR