4-(Dihydroxyborophenyl)acetylene 98%

Supplier: Ambeed
Warning

Synonyms: B-(4-Ethynylphenyl)boronic Acid, (4-Ethynylphenyl)boronic Acid, 4-Ethynylbenzeneboronic Acid

A418090-100MG A418090-1G A418090-250MG
77016-420EA 16.98 USD
77016-420 77016-422 77016-424
4-(Dihydroxyborophenyl)acetylene 98%
4-(Dihydroxyborophenyl)acetylene

Precaución: for Research Use Only

Formula: C₈H₇BO₂
MW: 145.95 g/mol
MDL Number: MFCD08689553
CAS Number: 263368-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR