1-Bromo-3-chloro-4-ethoxy-2-fluorobenzene 97%

Supplier: Ambeed
Warning

A417588-100MG A417588-1G A417588-250MG
76990-402EA 45.62 USD
76990-402 76990-404 76990-406
1-Bromo-3-chloro-4-ethoxy-2-fluorobenzene 97%
1-Bromo-3-chloro-4-ethoxy-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
MW: 253.50 g/mol
MDL Number: MFCD19440928
CAS Number: 909122-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR