Telaprevir 97%

Supplier: Ambeed
Warning

Synonyms: (1S,3aR,6aS)-2-((S)-2-((S)-2-Cyclohexyl-2-(pyrazine-2-carboxamido)acetamido)-3,3-dimethylbutanoyl)-N-((S)-1-(cyclopropylamino)-1,2-dioxohexan-3-yl)octahydrocyclopenta[c]pyrrole-1-carboxamide, (3S,3aS,6aR)-2-[(2S)-2-[[(2S)-2-cyclohexyl-2-(pyrazine-2-carbonylamino)acetyl]amino]-3,3-dimethylbutanoyl]-N-[(3S)-1-(cyclopropylamino)-1,2-dioxohexan-3-yl]-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-3-carboxamide

A417401-10MG A417401-1MG A417401-25MG A417401-50MG A417401-5MG
77033-814EA 84.88 USD
77033-814 77033-816 77033-818 77033-820 77033-822
Telaprevir 97%
Telaprevir

Precaución: for Research Use Only

Formula: C₃₆H₅₃N₇O₆
MW: 679.85 g/mol
MDL Number: MFCD11616089
CAS Number: 402957-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR