5-Bromo-1-(difluoromethyl)pyridin-2(1h)-one 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-(difluoromethyl)pyridin-2(1H)-one

A417241-1G A417241-5G
77033-810EA 264.17 USD
77033-810 77033-812
5-Bromo-1-(difluoromethyl)pyridin-2(1h)-one 95%
5-Bromo-1-(difluoromethyl)pyridin-2(1h)-one

Precaución: for Research Use Only

Formula: C₆H₄BrF₂NO
MW: 224.003 g/mol
MDL Number: MFCD12911848
CAS Number: 832735-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR