7-Iodo-6-methoxy-4-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A417129-100MG A417129-250MG
77016-414EA 49.87 USD
77016-414 77016-416
7-Iodo-6-methoxy-4-methyl-1H-indazole 97%
7-Iodo-6-methoxy-4-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉IN₂O
MW: 288.09 g/mol
MDL Number: MFCD20923508
CAS Number: 1352397-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR