3-Chloro-6-(methylsulfonyl)pyridazine 95%
Supplier: Ambeed
|
Warning
|
A416883-250MG
A416883-1G
77033-798EA
64.72
USD
77033-798
77601-808
3-Chloro-6-(methylsulfonyl)pyridazine 95%
3-Chloro-6-(methylsulfonyl)pyridazine
Precaución: for Research Use Only
Formula:
C5H5ClN2O2S MW: 192.6234 g/mol |
MDL Number:
MFCD03931474 CAS Number: 7145-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...