4'-Bromo-[1,1'-biphenyl]-2,4-diol 95%
Supplier: Ambeed
|
Warning
|
A415822-100MG
A415822-1G
A415822-250MG
77016-402EA
24
USD
77016-402
77016-404
77016-406
4'-Bromo-[1,1'-biphenyl]-2,4-diol 95%
4'-Bromo-[1,1'-biphenyl]-2,4-diol
Precaución: for Research Use Only
Formula:
C₁₂H₉BrO₂ |
MDL Number:
MFCD23381094 CAS Number: 1418117-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...