1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct 95%

Supplier: Ambeed
Danger

A415779-1G A415779-25G A415779-5G
76990-392EA 10 USD
76990-392 76990-394 76990-396
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct 95%
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₄S₂
MDL Number: MFCD22377802
CAS Number: 119752-83-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR