Quinoxaline-5-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: Quinoxaline-5-carbaldehyde, 5-Formylquinoxaline

A415664-100MG A415664-1G A415664-250MG
77016-396EA 47.75 USD
77016-396 77016-398 77016-400
Quinoxaline-5-carboxaldehyde 95%
Quinoxaline-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆N₂O
MW: 158.157 g/mol
MDL Number: MFCD00805833
CAS Number: 141234-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR