Disodium hexahydroxoplatinate 98%

Supplier: Ambeed
Warning

Synonyms: Disodium platinate, Sodium hexahydroxyplatinate(IV)

A415302-100MG A415302-1G A415302-250MG
76990-384EA 44.56 USD
76990-384 76990-386 76990-388
Disodium hexahydroxoplatinate 98%
Disodium hexahydroxoplatinate

Precaución: for Research Use Only

Formula: H₆Na₂O₆Pt
MW: 343.1 g/mol
MDL Number: MFCD00014236
CAS Number: 12325-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR