(S)-3-Aminobutan-2-one hydrochloride 95%

Supplier: Ambeed
Danger

A414729-1G A414729-250MG
77016-392EA 471.04 USD
77016-392 77016-394
(S)-3-Aminobutan-2-one hydrochloride 95%
(S)-3-Aminobutan-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD16619120
CAS Number: 54680-72-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR