2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-pyrrolidinobenzoic Acid, 2-Chloro-4-(pyrrolidin-1-yl)benzoic acid

A414556-1G A414556-250MG A414556-5G A414556-100MG
77033-774EA 171.87 USD
77033-774 77033-776 77033-778 77673-014
2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid 97%
2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO₂
MW: 225.68 g/mol
MDL Number: MFCD03001210
CAS Number: 192513-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR