Ethyl 4-bromo-3-fluorobenzoate 96%

Supplier: Ambeed
Warning

A414470-100G A414470-1G A414470-25G A414470-5G
76990-374EA 199 USD
76990-374 76990-376 76990-378 76990-380
Ethyl 4-bromo-3-fluorobenzoate 96%
Ethyl 4-bromo-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD09751983
CAS Number: 1130165-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR