(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol 90%

Supplier: Ambeed
Warning

Synonyms: rel-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol, BCN-OH

A413936-250MG A413936-1G A413936-5G
77016-390EA 401.03 USD
77016-390 77568-834 77572-288
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol 90%
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol

Precaución: For research use only.

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD26142970
CAS Number: 1263166-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR