4-Bromo-1-(difluoromethyl)-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
A413483-100MG
A413483-250MG
A413483-1G
76990-362EA
32
USD
76990-362
76990-364
77890-850
4-Bromo-1-(difluoromethyl)-1H-pyrazole 97%
4-Bromo-1-(difluoromethyl)-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₄H₃BrF₂N₂ |
MDL Number:
MFCD11558132 CAS Number: 956477-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...