4-Bromo-2-chloro-1-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A413382-1G A413382-25G
76990-358EA 20.16 USD
76990-358 76990-360
4-Bromo-2-chloro-1-(trifluoromethoxy)benzene 98%
4-Bromo-2-chloro-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MW: 275.4503 g/mol
MDL Number: MFCD10699114
CAS Number: 158579-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR