Methyl 2-chloroquinoline-6-carboxylate 98%

Supplier: Ambeed
Warning

A413112-100MG A413112-1G A413112-5G A413112-250MG
77033-762EA 18 USD
77033-762 77033-764 77033-766 77601-804
Methyl 2-chloroquinoline-6-carboxylate 98%
Methyl 2-chloroquinoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO₂
MDL Number: MFCD17215626
CAS Number: 849807-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR