4-(Diisopropylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Diisopropylcarbamoyl)phenyl)boronic acid, 4-(N,N-Diisopropylaminocarbonyl)phenylboronic acid, 4-(N,N-Diisopropylaminocarbonyl)benzeneboronic acid

A412775-1G A412775-250MG A412775-5G
77016-384EA 70 USD
77016-384 77016-386 77016-388
4-(Diisopropylcarbamoyl)phenylboronic acid 98%
4-(Diisopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
MDL Number: MFCD06659884
CAS Number: 850568-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR