2,6-Dibromo-4-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A412404-100MG A412404-10G A412404-1G A412404-250MG A412404-5G
76990-336EA 27 USD
76990-336 76990-338 76990-340 76990-342 76990-344
2,6-Dibromo-4-(trifluoromethyl)pyridine 95%
2,6-Dibromo-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₃N
MDL Number: MFCD22682722
CAS Number: 1000152-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR