2-Chloro-4-isopropylpyrimidine 98%

Supplier: Ambeed
Danger

A412316-100MG A412316-250MG
76990-332EA 22 USD
76990-332 76990-334
2-Chloro-4-isopropylpyrimidine 98%
2-Chloro-4-isopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MDL Number: MFCD09475913
CAS Number: 941294-36-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR