3-Methylpyrrolidin-3-ol 95%

Supplier: Ambeed
Danger

A412126-1G A412126-250MG A412126-5G
77033-754EA 26.53 USD
77033-754 77033-756 77033-758
3-Methylpyrrolidin-3-ol 95%
3-Methylpyrrolidin-3-ol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.147 g/mol
MDL Number: MFCD11100984
CAS Number: 125032-87-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR