Fmoc-Aph(Hor)-OH 98% (mix 2% EA)

Supplier: Ambeed
Warning

A411657-100MG A411657-1G A411657-250MG
77016-378EA 29 USD
77016-378 77016-380 77016-382
Fmoc-Aph(Hor)-OH 98% (mix 2% EA)
Fmoc-Aph(Hor)-OH

Precaución: for Research Use Only

Formula: C₂₉H₂₆N₄O₇
MDL Number: MFCD27923015
CAS Number: 1253282-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR