3,5-Bis(trifluoromethyl)benzamide 97%

Supplier: Ambeed
Warning

A411347-100G A411347-1G A411347-25G A411347-5G
77004-046EA 197.33 USD
77004-046 77004-048 77004-050 77004-052
3,5-Bis(trifluoromethyl)benzamide 97%
3,5-Bis(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₉H₅F₆NO
MW: 257.14 g/mol
MDL Number: MFCD00015511
CAS Number: 22227-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR