(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone 95%

Supplier: Ambeed
Warning

Synonyms: (2R,3S)-(-)-2,3-Diphenyl-6-oxomorpholine, N-BOC protected, (2R,3S)-tert-Butyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate

A411309-1G A411309-250MG A411309-5G
77004-042EA 33.95 USD
77004-042 77004-044 77594-254
(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone 95%
(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
MDL Number: MFCD00074955
CAS Number: 112741-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR