(2S,3R,4S,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol 95%

Supplier: Ambeed
Warning

A411294-100G A411294-1G A411294-250MG A411294-25G A411294-5G
77016-368EA 531 USD
77016-368 77016-370 77016-372 77016-374 77016-376
(2S,3R,4S,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol 95%
(2S,3R,4S,5S,6R)-2-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₂₂H₂₇ClO₇
MDL Number: MFCD27923102
CAS Number: 714269-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR