3-[(Ethylamino)methyl]phenol 95%

Supplier: Ambeed
Warning

A411263-1G A411263-5G
77016-364EA 41.38 USD
77016-364 77016-366
3-[(Ethylamino)methyl]phenol 95%
3-[(Ethylamino)methyl]phenol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.206 g/mol
MDL Number: MFCD09931765
CAS Number: 91239-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR