(R)-2-Amino-3-(2-nitrophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Phe(2-NO₂)-OH

A411159-1G A411159-250MG A411159-5G
77004-036EA 115.64 USD
77004-036 77004-038 77004-040
(R)-2-Amino-3-(2-nitrophenyl)propanoic acid 98%
(R)-2-Amino-3-(2-nitrophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.18 g/mol
MDL Number: MFCD00270361
CAS Number: 169383-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR