2-(Phenylazo)malononitrile 96%

Supplier: Ambeed
Warning

A410746-100G A410746-10G A410746-25G A410746-5G
77033-746EA 66.84 USD
77033-746 77033-748 77033-750 77033-752
2-(Phenylazo)malononitrile 96%
2-(Phenylazo)malononitrile

Precaución: for Research Use Only

Formula: C₉H₆N₄
MW: 170.17 g/mol
MDL Number: MFCD00524838
CAS Number: 6017-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR