Methyl 3-Bromo-1-methylindole-6-carboxylate 95%

Supplier: Ambeed
Warning

A410188-250MG A410188-5G
77033-742EA 21.22 USD
77033-742 77033-744
Methyl 3-Bromo-1-methylindole-6-carboxylate 95%
Methyl 3-Bromo-1-methylindole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
MDL Number: MFCD12828701
CAS Number: 1186663-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR