(4-(4-Fluorophenoxy)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A409889-5G A409889-1G A409889-250MG
77033-740EA 310 USD
77033-740 77872-650 77872-651
(4-(4-Fluorophenoxy)phenyl)boronic acid 95%
(4-(4-Fluorophenoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₃
MDL Number: MFCD13182406
CAS Number: 361437-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR