Methyl-5-bromo-2-chloropyridine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-bromo-2-chloroisonicotinate
A409706-1G
A409706-250MG
A409706-25G
A409706-5G
77033-732EA
35.01
USD
77033-732
77033-734
77033-736
77033-738
Methyl-5-bromo-2-chloropyridine-4-carboxylate 97%
Methyl-5-bromo-2-chloropyridine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₅BrClNO₂ MW: 250.49 g/mol |
MDL Number:
MFCD07375113 CAS Number: 886365-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...