2-[2-(Dimethylamino)ethoxy]ethanol 98%

Supplier: Ambeed
Danger

Synonyms: Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether, 2-(2-(Dimethylamino)ethoxy)ethanol

A409648-10G A409648-25G
77004-028EA 6.37 USD
77004-028 77594-244
2-[2-(Dimethylamino)ethoxy]ethanol 98%
2-[2-(Dimethylamino)ethoxy]ethanol

Precaución: for Research Use Only

Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
MDL Number: MFCD00059602
CAS Number: 1704-62-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR