2-Methylthiazole-5-carbonitrile 95%

Supplier: Ambeed
Danger

A409472-100MG A409472-1G A409472-250MG
76990-316EA 34 USD
76990-316 76990-318 76990-320
2-Methylthiazole-5-carbonitrile 95%
2-Methylthiazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₂S
MW: 124.16 g/mol
MDL Number: MFCD11506340
CAS Number: 65735-10-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR